Learn More
Rabies Virus Glycoprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 398.00€
Specifications
| Antigen | Rabies Virus Glycoprotein |
|---|---|
| Dilution | Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281404
|
Novus Biologicals
NBP3-43766-100ul |
100 μL |
398.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281405
|
Novus Biologicals
NBP3-43766-20ul |
20 μL |
201.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rabies Virus Glycoprotein Polyclonal antibody specifically detects Rabies Virus Glycoprotein in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Rabies Virus Glycoprotein | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Produced in rabbits immunized with purified, recombinant Rabies Virus Glycoprotein | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |