missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rabenosyn 5 Polyclonal specifically detects Rabenosyn 5 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Rabenosyn 5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | 110 kDa protein, FLJ34993, FYVE finger-containing Rab5 effector protein rabenosyn-5, FYVE-finger-containing Rab5 effector protein rabenosyn-5, MGC126210, Rabenosyn-5, ZFYVE20, Zinc finger FYVE domain-containing protein 20, zinc finger, FYVE domain containing 20 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human Rabenosyn 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?