missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Rabbit Multiclonal Antibody, Anti-NOTCH3, RM1102, Carrier Free, 100 μg
Description
- Host Species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Applications: WB, IHC-P, Flow Cyt (Intra)
- Reactivity: Human
- Purification: Affinity purification Protein A
- Form: Liquid
- Buffer: PBS, pH 7.2 to 7.4
- Features: Carrier free, conjugation ready
Specifications
Specifications
| Buffer | PBS, pH 7.2 to 7.4 |
| Detection Location | In-Blot Detection, In-Cell Detection |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Target Molecule | NOTCH3 |
| Cross Reactivity | Human |
| Formulation | PBS, pH 7.2 to 7.4 |
| Quantity | 100 μg |
| For Use With (Application) | Flow Cytometry, IHC-P, Western Blot |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?