Learn More
Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems VC003-025
 View more versions of this product
Product Code. 18792418
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Rabbit IgG
25 mL
Secondary
Detects heavy ( gamma ) chains on rabbit IgG and light chains on all rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin rabbit serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
6 months from date of receipt, 2 to 8 degreesC as supplied.
Immunohistochemistry
HRP Polymer
Supplied as a solution in PBS with preservative.
Goat
Affinity Purified
RUO
3500