Learn More
SNCA (phospho Y133), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of SNCA.
Brand: Abnova PAB25866.100ug
Description
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Four alternatively spliced transcripts encoding two different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: E-G-YpQ-DSpecifications
| SNCA | |
| Polyclonal | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| SNCA | |
| SNCA | |
| Synthetic phosphopeptide corresponding to residues surrounding Y133 of human SNCA. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of SNCA. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MGC110988/NACP/PARK1/PARK4/PD1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6622 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only