Learn More
SLITRK4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLITRK4.
Brand: Abnova PAB13338.100ug
Description
Members of the SLITRK family, such as SLITRK4, are integral membrane proteins with 2 N-terminal leucine-rich repeat (LRR) domains similar to those of SLIT proteins (see SLIT1; MIM 603742). Most SLITRKs, including SLITRK4, also have C-terminal regions that share homology with neurotrophin receptors (see NTRK1; MIM 191315). SLITRKs are expressed predominantly in neural tissues and have neurite-modulating activity (Aruga et al., 2003 [PubMed 14557068]).[supplied by OMIM
Specifications
| SLITRK4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLITRK4. | |
| In PBS (0.02% sodium azide) | |
| DKFZp547M2010 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLITRK4 | |
| SLITRK4 | |
| A synthetic peptide corresponding to C-terminus 13 amino acids of human SLITRK4. | |
| RUO | |
| 139065 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |