missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Sheep IgM (H+L) Secondary Antibody, DyLight 650, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgM (H+L) Polyclonal specifically detects IgM (H+L) in Sheep samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Ovine IgM |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?