missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Sheep IgA Secondary Antibody, DyLight 755, Novus Biologicals™

Product Code. 30228003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228003 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30228003 Supplier Novus Biologicals Supplier No. NBP268361IR
  • ONLINE EXCLUSIVE PRICE
    628.00€
    593.25€ / 1mL
    Save 34.75€
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgA Polyclonal specifically detects IgA in Sheep samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Immunoglobulin A
Host Species Rabbit
Immunogen Ovine IgA
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.