Learn More
SEC16B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of SEC16B.
Brand: Abnova PAB19201.100ug
Description
SEC16B is a mammalian homolog of S. cerevisiae Sec16 that is required for organization of transitional endoplasmic reticulum (ER) sites and protein export (Bhattacharyya and Glick, 2007 [PubMed 17192411]).[supplied by OMIM
Specifications
| SEC16B | |
| Polyclonal | |
| Unconjugated | |
| Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| SEC16B | |
| DKFZp686C2486/FLJ23871/FLJ25761/FLJ33652/FLJ36620/LZTR2/PGPR-p117/RGPR/SEC16S | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 89866 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of SEC16B. | |
| In PBS (0.02% sodium azide) | |
| NP_149118 | |
| SEC16B | |
| A synthetic peptide corresponding to 18 amino acids near C-terminus of human SEC16B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only