missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S14G-in, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of S14G-in.
Brand: Abnova PAB8508.25ug
This item is not returnable.
View return policy
Specifications
| S14G-in | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of S14G-in. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| A synthetic peptide corresponding to human S14G-Humanin. | |
| RUO | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 25 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction