missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Rat IgG Fc Secondary Antibody, PE/Cy5.5, R&D Systems™

Product Code. 18603699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18603699 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18603699 Supplier Novus Biologicals Supplier No. NBP260687PECY55
  • ONLINE EXCLUSIVE PRICE
    783.00€
    740.25€ / 500µL
    Save 42.75€
    5% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgG Fc Polyclonal antibody specifically detects IgG Fc in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate PE-Cyanine5.5
Formulation PBS
Host Species Rabbit
Immunogen Rat IgG F(c) fragment
Purification Method Multi-step
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.