Learn More
NOS2 (phospho Y151), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NOS2.
Brand: Abnova PAB29225.100uL
Description
Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. This gene encodes a nitric oxide synthase which is expressed in liver and is inducible by a combination of lipopolysaccharide and certain cytokines. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Specifications
| NOS2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NOS2. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| HEP-NOS/INOS/NOS/NOS2A | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4843 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| NOS2 | |
| NOS2 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y151 of human NOS2. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of NOS2 only when phosphorylated at tyrosine 151. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only