missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nephrophthisis (pic), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Nephrophthisis (pic).
Brand: Abnova PAB10408.250ug
This item is not returnable.
View return policy
Specifications
| Nephrophthisis (pic) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Nephrophthisis (pic). | |
| In PBS (0.08% sodium azide) | |
| Recombinant GST fusion protein corresponding to amino acid 612-691 of mouse pic. | |
| RUO | |
| Mouse | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western blot (2 to 10 ug/mL) Immunoprecipitation (10 to 20 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 250 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction