missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgM whole molecule,Polyclonal Secondary Antibody, Abnova™
Specifications
Specifications
| Antigen | Rabbit anti-Mouse IgM whole molecule |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-mouse IgG recognizes mouse IgM whole molecule. This secondary antibody was purified using delipidation and defibrination. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM sodium phosphate, 140mM NaCl, pH 7.4 (0.01% (w/v) sodium azide) |
| Host Species | Rabbit |
| Immunogen | Mouse IgM whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against mouse IgM and mouse serum. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?