missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG2b Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Product Code. 18262443 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18262443 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18262443 Supplier Novus Biologicals Supplier No. NBP172674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG2b Heavy Chain Polyclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2b Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:1000 - 1:3000, ELISA 1:8000 - 1:32000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:50 - 1:200, Dot Blot
Host Species Rabbit
Immunogen Mouse IgG2b heavy chain
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Specificity was confirmed by ELISA at less than 1% cross reactivity against other mouse heavy or light chain isotypes.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.