missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG2a Heavy Chain Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72794JF549
This item is not returnable.
View return policy
Description
IgG2a Heavy Chain Polyclonal antibody specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot, Fluorophore-linked immunosorbent assay
Specifications
| IgG2a Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Mouse IgG2a heavy chain | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot | |
| Janelia Fluor 549 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction