missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG1 Secondary Antibody (20) [FITC], Novus Biologicals™

Product Code. 30281474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30281474 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281474 Supplier Novus Biologicals Supplier No. NBP343768F
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Applications ELISA
Classification Monoclonal
Clone 20
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Rabbit
Immunogen Recombinant Mouse IgG1 Fc Protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.