missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72672
369.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG1 Heavy Chain | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Specificity was confirmed by ELISA at less than 1% cross reactivity against other mouse heavy or light chain isotypes. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2500, ELISA 1:5000, Immunohistochemistry 1:200 - 1:1000, Dot Blot | |
| Mouse IgG1 heavy chain | |
| 1 mg | |
| Secondary | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction