missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
297.00€
Specifications
| Antigen | IgG, Whole Molecule |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Host Species | Rabbit |
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Mouse
Specifications
| IgG, Whole Molecule | |
| Unconjugated | |
| Rabbit | |
| Secondary |
| Polyclonal | |
| Purified | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title