missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™

Product Code. 18464297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18464297 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18464297 Supplier Novus Biologicals Supplier No. NBP260654UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Conjugate DyLight 350
Dilution Flow Cytometry : 1:20-1:2000, Immunomicroscopy : 1:20-1:2000
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified mouse IgG (H&L).
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.