missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Monkey IgA Fc Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69199
This item is not returnable.
View return policy
Description
IgA Fc Polyclonal antibody specifically detects IgA Fc in Monkey samples. It is validated for Immunoelectrophoresis
Specifications
| IgA Fc | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunoelectrophoresis | |
| LYOPH | |
| Whole antiserum | |
| Pools of purified IgA isolated from Rhesus monkey serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Monkey | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur