Learn More
MMP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MMP9.
Brand: Abnova PAB12714.100ug
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. [provided by RefSeq
Specifications
| MMP9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MMP9. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CLG4B/GELB/MMP-9 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼92 KDa of human MMP9 protein. | |
| Store at 4°C. For long term storage store at -20°C to -80°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MMP9 | |
| MMP9 | |
| A synthetic peptide corresponding to the hinge region of human MMP9. | |
| RUO | |
| 4318 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only