Learn More
MAP1LC3A Rabbit anti-Bovine, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against MAP1LC3A.
Brand: Abnova PAB15521.100uL
Description
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| MAP1LC3A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against MAP1LC3A. | |
| In PBS (50% glycerol, 0.1% BSA, 0.05% sodium azide) | |
| LC3/LC3A/MAP1ALC3/MAP1BLC3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Immunogen has 100% homology to rat and 97% homology to bovine. This antibody is useful for Western blot, where a band is seen ∼14 KDa. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:2500) The optimal working dilution should be determined by the end user. | |
| MAP1LC3A | |
| MAP1LC3A | |
| N-terminus of human MAP1LC3A. | |
| RUO | |
| 84557 | |
| Bovine, Human, Mouse, Rat, Zebrafish | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only