missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Llama IgG (H+L) Secondary Antibody, PE/Cy5.5, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-75091PECY55
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoassay
Specifications
| IgG (H+L) | |
| Polyclonal | |
| PBS | |
| This Rabbit anti-Llama IgG (H+L) Secondary Antibody was developed against purified llama IgG (H&L). | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Llama | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoassay | |
| PE-Cyanine5.5 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction