missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Llama IgG (H+L) Secondary Antibody, DyLight 488, R&D Systems™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot : 1:30000 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Purified Llama IgG |
| Purification Method | Affinity purified |
| Quantity | 0.5 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?