missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH (Keyhole Limpet Hemocyanin), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against KLH (Keyhole Limpet Hemocyanin).
Brand: Abnova PAB10347.1mg
This item is not returnable.
View return policy
Specifications
| KLH (Keyhole Limpet Hemocyanin) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against KLH (Keyhole Limpet Hemocyanin). | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Keyhole Limpet Hemocyanin, KLH. | |
| RUO | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:400000) Western Blot (1:40000) Immunohistochemistry (1:20000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mg | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction