Learn More
KAT5 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KAT5.
Brand: Abnova PAB18305.100ug
Description
The protein encoded by this gene belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein. HATs play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. This protein is a histone acetylase that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Specifications
| KAT5 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| KAT5 | |
| KAT5 | |
| A synthetic peptide corresponding to human KAT5. | |
| 100 μg | |
| Primary | |
| This antibody is specific to KAT5. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of KAT5. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| ESA1/HTATIP/HTATIP1/PLIP/TIP/TIP60/cPLA2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 10524 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only