missing translation for 'onlineSavingsMsg'
Learn More

Mycobacterium tuberculosis EsaT-6 (Rv3875), Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16000037
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16000037 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16000037 Supplier Abnova Supplier No. PAB1266.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against Mycobacterium tuberculosis EsaT-6 (Rv3875).

Specifications

Antigen Mycobacterium tuberculosis EsaT-6 (Rv3875)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Mycobacterium tuberculosis EsaT-6 (Rv3875).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Rabbit
Immunogen ESAT-6 (Rv3875) of Mycobacterium tuberculosis.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with EsaT-6, a 11.8 KDa antigen encoded by gene Rv3875 of Mycobacterium tuberculosis. This antibody recognizes EsaT-6 of Mycobacterium tuberculosis H37Rv and Mycobacterium bovis; it reacts also with recombinant antigen produced in Escherichia coli.
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.