missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Horse IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™
Specifications
Specifications
| Antigen | Rabbit anti-Horse IgG F(ab')2 fragment |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-horse IgG recognizes horse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In 10mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) |
| Host Species | Rabbit |
| Immunogen | Horse IgG F(ab')2 fragment |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?