Learn More
GSK3B (phospho S9), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B.
Brand:  Abnova PAB10055.100ug
Product Code. 16049026
  • 870.00€ / 100µg
Description
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifications
Specifications
GSK3B
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
NM_002093(Human);P49841(Human)
Rabbit
100 μg
Primary
This phospho specific polyclonal antibody reacts with phosphorylated pS9 of human GSK3B. Reactivity with non-phosphorylated human GSK3B is minimal.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000-1:50000) Western Blot (1:500-1:3000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
GSK3B
GSK3B
Synthetic phosphopeptide corresponding to residues surrounding S9 of human GSK3B.
RUO
2932
Canine, Chicken, Frog, Human, Rat