Learn More
GPR98 Rabbit anti-Horse, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR98.
Brand: Abnova PAB26383.50ug
Description
This gene encodes a member of the G-protein coupled receptor superfamily. The encoded protein contains a 7-transmembrane receptor domain, binds calcium and is expressed in the central nervous system. Mutations in this gene are associated with Usher syndrome 2 and familial febrile seizures. Several alternatively spliced transcripts have been described. [provided by RefSeq
Specifications
| GPR98 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR98. | |
| In PBS (0.09% sodium azide) | |
| Q8WXG9 | |
| GPR98 | |
| A synthetic peptide corresponding to 19 amino acids at N-terminal extracellular domain of human GPR98. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10-26 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR98 | |
| DKFZp761P0710/FEB4/KIAA0686/MASS1/USH2B/USH2C/VLGR1/VLGR1b | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 84059 | |
| Equine, Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only