missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Golden Syrian hamster IgG whole molecule,Polyclonal Secondary Antibody, Abnova™

Product Code. 16091528
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16091528 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16091528 Supplier Abnova Supplier No. PAB9235.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Antigen Rabbit anti-Golden Syrian hamster IgG whole molecule
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit anti-Golden Syrian hamster IgG recognizes Golden Syrian hamster IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Golden Syrian hamster IgG whole molecule
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation.
Quantity 1 mg
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-rabbit IgG F(c). Limited reactivity will occur with Armenian hamster IgG.
Target Species Hamster
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.