missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Golden Syrian hamster IgG F(ab')2 fragment, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10609.1mg
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Golden Syrian hamster IgG F(ab')2 fragment | |
| Polyclonal | |
| Rabbit anti-Golden Syrian hamster IgG recognizes Golden Syrian hamster IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Golden Syrian hamster IgG F(ab')2 fragment | |
| 1 mg | |
| No reaction was observed against hamster IgG F(c). | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence | |
| Texas Red | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Hamster | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction