missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat Light Chain Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69509
This item is not returnable.
View return policy
Description
Light Chain Polyclonal antibody specifically detects Light Chain in Goat samples. It is validated for Immunoelectrophoresis
Specifications
| Light Chain | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Reconstitute by adding 1 mL sterile distilled water. Working dilutions are prepared by adding sterile phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunoelectrophoresis | |
| LYOPH | |
| Whole antiserum | |
| Purified light chains isolated from polyclonal goat IgG. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Goat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido