Learn More
Rabbit anti-Goat IgM (H+L) Secondary Antibody, R&D Systems™
Product Code. 18695787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18695787 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18695787 Supplier Novus Biologicals Supplier No. NBP269654
  • ONLINE EXCLUSIVE PRICE
    573.00€
    541.80€ / 10mg
    Save 31.20€
    5% Off
 Restricted Item
Estimated Shipment: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Goat samples. It is validated for Western Blot, ELISA, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM (H+L)
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Lyophilized from PBS (pH 7.2)
Host Species Rabbit
Immunogen Purified normal IgM isolated from pooled goat serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Goat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Mehr anzeigen Weniger anzeigen