missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Texas Red |
| Dilution | Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free |
| Host Species | Rabbit |
| Immunogen | Goat IgG whole molecule |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?