missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72833
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| DyLight 680 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 100 ul deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000 | |
| Goat IgG, whole molecule | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody will react with heavy chains of Goat IgG and with light chains of most Goat immunoglobulins. | |
| Goat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction