Learn More
Rabbit anti-Goat IgG F(ab')2 Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-73467
Product Code. 18285313
  • 406.00€ / 2mg
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG F(ab')2
Polyclonal
HRP
immunoglobulin heavy constant gamma 1 (G1m marker)
Goat IgG F(ab')2 fragment
2 mg
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Dot Blot
LYOPH
Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500, Dot Blot
Rabbit
Affinity Purified
RUO
No reaction was observed against Goat IgG F(c).
Goat