Learn More
FYN (phospho Y530), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist. [provided by RefSeq
Specifications
Specifications
| Antigen | FYN |
| Applications | Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of FYN. |
| Dilution | Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. |
| Formulation | In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) |
| Gene | FYN |
| Gene Accession No. | P06241 |
| Gene Alias | MGC45350/SLK/SYN |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.