missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Feline IgG (H+L) Secondary Antibody, Novus Biologicals™

Product Code. 18599863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
2 mg
Unit Size:
0.5mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18599863 2 mg 2mg
18538782 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18599863 Supplier Novus Biologicals Supplier No. NBP174767
  • ONLINE EXCLUSIVE PRICE
    298.00€
    281.40€ / 2mg
    Save 16.60€
    5% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA)
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Host Species Rabbit
Immunogen This Rabbit anti-Feline IgG (H+L) Secondary Antibody was developed against purified feline IgG (H&L).
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Feline
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.