missing translation for 'onlineSavingsMsg'
Learn More

Rabbit Anti-Donkey IgG H&L (Texas Red ™), Abcam

Product Code. 32116404
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116404 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116404 Supplier Abcam Supplier No. ab67631mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rabbit Anti-Donkey IgG H&L (Texas Red ™)

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Donkey IgG
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Immunohistochemistry (Frozen), Immunomicroscopy
Classification Polyclonal
Conjugate Texas Red
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Host Species Rabbit
Immunogen Donkey IgG
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Donkey
Content And Storage -20°C
Product Type Primary Antibody
Form Lyophilized
Isotype IgG
Shipping Condition Blue Ice
Target Molecule Donkey IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.