missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Dog IgG whole molecule, Rhodamine, TRITC, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10534.2mg
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Dog IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-dog IgG recognizes dog IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Dog IgG whole molecule | |
| 2 mg | |
| Dog | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence | |
| Rhodamine | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction