missing translation for 'onlineSavingsMsg'
Learn More

DCLRE1C (phospho S645), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16162090
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16162090 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16162090 Supplier Abnova Supplier No. PAB15904.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of DCLRE1C.

This gene encodes a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5'-3' exonuclease activity; it also exhibits endonuclease activity on 5' and 3' overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide. Mutations in this gene cause Athabascan-type severe combined immunodeficiency (SCIDA). [provided by RefSeq]

Specifications

Antigen DCLRE1C
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of DCLRE1C.
Dilution ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene DCLRE1C
Gene Alias A-SCID/DCLREC1C/FLJ11360/FLJ36438/RS-SCID/SCIDA/SNM1C
Gene Symbols DCLRE1C
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S645 of human DCLRE1C.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64421
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.