missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Chicken IgG Fc Secondary Antibody, PE/Cy7, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62593PECY7
627.30 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG Fc | |
| Polyclonal | |
| PBS | |
| Chicken IgG F(c) fragment | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Chicken | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| PE-Cyanine7 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction