Learn More
BLM Rabbit anti-Human, Polyclonal Antibody, Abnova™
Description
The Bloom syndrome gene product is related to the RecQ subset of DExH box-containing DNA helicases and has both DNA-stimulated ATPase and ATP-dependent DNA helicase activities. Mutations causing Bloom syndrome delete or alter helicase motifs and may disable the 3'-5' helicase activity. The normal protein may act to suppress inappropriate recombination. [provided by RefSeq]
Specifications
Specifications
| Antigen | BLM |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of BLM. |
| Dilution | Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gene | BLM |
| Gene Accession No. | P54132 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.