Learn More
APOBEC3G Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APOBEC3G.
Brand: Abnova PAB2470.400uL
Description
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. The protein encoded by this gene has been found to be a specific inhibitor of human immunodeficiency virus-1 (HIV-1) infectivity. [provided by RefSeq
Specifications
| APOBEC3G | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of APOBEC3G. | |
| In PBS (0.09% sodium azide) | |
| AAG14956 | |
| APOBEC3G | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human APOBEC3G. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| APOBEC3G | |
| ARP9/CEM15/FLJ12740/MDS019/bK150C2.7/dJ494G10.1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 60489 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only