missing translation for 'onlineSavingsMsg'
Learn More

Aof 2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16013127
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16013127 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16013127 Supplier Abnova Supplier No. PAB1844.400uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of Aof 2.

This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen Aof 2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Aof 2.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene AOF2
Gene Accession No. NP_055828;O60341
Gene Alias BHC110/KDM1/KIAA0601/LSD1
Gene Symbols AOF2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of human AOF2.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23028
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.