Learn More
ALDH6A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDH6A1.
Brand: Abnova PAB3127.400uL
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids. [provided by RefSeq
Specifications
| ALDH6A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ALDH6A1. | |
| In PBS (0.09% sodium azide) | |
| NP_005580;Q02252 | |
| ALDH6A1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ALDH6A1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. | |
| ALDH6A1 | |
| MGC40271/MMSADHA/MMSDH | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 4329 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only