Learn More
AKT2 (phospho S474), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT2.
Brand: Abnova PAB8183.100uL
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Specifications
| AKT2 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| AKT2 | |
| AKT2 | |
| Synthetic phosphopeptide corresponding to residues surrounding S474 of human AKT2. | |
| 100 μL | |
| Primary | |
| Akt2 (phospho-Ser474) antibody detects endogenous levels of Akt2 only when phosphorylated at serine 474. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT2. | |
| In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PKBB/PKBBETA/PRKBB/RAC-BETA | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 208 | |
| Human, Mouse, Rat | |
| Liquid |