missing translation for 'onlineSavingsMsg'
Learn More

Actin (32kDa fragment), Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16032487
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16032487 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16032487 Supplier Abnova Supplier No. PAB16937.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of Actin (32kDa fragment).

Specifications

Antigen Actin (32 KDa fragment)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Actin (32 KDa fragment).
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation No additive
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of 32 kDa actin fragment.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody has been directly tested for reactivity in human and rat. specific to the ∼ 32 KDa fractin protein in Western blots with no reactivity to intact actin. There is often a ladder of smaller bands in cells or culture or in vivo preparations due to further degradation by other proteases.
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.